Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK007911.1 | - | 1 - 2 | 80 | 79 | |
AK148544.1 | - | 1 - 2 | 82 | 78 | ||
BC012975.1 | - | 1 - 2 | 83 | 4218718 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |