Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205369.1 | NP_001192298.1 | 8 - 9 | 1302 | 81 | |
NM_001205371.1 | NP_001192300.1 | 8 - 9 | 892 | 81 | ||
NM_177054.5 | NP_796028.2 | 7 - 8 | 1203 | 81 | ||
GenBank mRNA | AK131751.1 | - | 8 - 9 | 892 | 81 | |
AY484583.1 | - | 7 - 8 | 1043 | 81 | ||
BC064446.1 | - | 8 - 9 | 1302 | 6843743 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |