Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081238.2 | NP_001074707.1 | 7 - 8 | 2685 | 78 | |
NM_001289842.1 | NP_001276771.1 | 7 - 8 | 2685 | 78 | ||
NM_001289843.1 | NP_001276772.1 | 6 - 7 | 2591 | 78 | ||
NM_001289844.1 | NP_001276773.1 | 7 - 8 | 1596 | 78 | ||
NM_001289845.1 | NP_001276774.1 | 4 - 5 | 525 | 78 | ||
GenBank mRNA | AK154227.1 | - | 7 - 8 | 78 | ||
AK156701.1 | - | 7 - 8 | 2604 | 78 | ||
AK157382.1 | - | 7 - 8 | 2604 | 78 | ||
BC044748.2 | - | 3 - 4 | 290 | 78 | ||
BC055302.1 | - | 4 - 5 | 488 | 78 | ||
BC066139.1 | - | 6 - 7 | 2533 | 78 | ||
BC066140.1 | - | 7 - 8 | 1494 | 30459312 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |