Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198029.2 | NP_932146.2 | 3 - 4 | 1117 | 145 | |
GenBank mRNA | AK030947.1 | - | 3 - 4 | 710 | 145 | |
AK050804.1 | - | 2 - 3 | 265 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |