Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128103.2 | NP_001121575.1 | 7 - 8 | 876 | 109 | |
GenBank mRNA | AK045930.1 | - | 7 - 8 | 877 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |