Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199113.1 | NP_001186042.1 | 2 - 3 | 188 | 90 | |
GenBank mRNA | AK180131.1 | - | 2 - 3 | 125 | 90 | |
AK191284.1 | - | 2 - 3 | 126 | 90 | ||
AK197634.1 | - | 2 - 3 | 125 | 90 | ||
AK205425.1 | - | 2 - 3 | 125 | 90 | ||
AK205461.1 | - | 2 - 3 | 125 | 90 | ||
BC004828.1 | - | 2 - 3 | 151 | 2631728 | 90 | |
BY773758.1 | - | 1 - 2 | 311 | 90 | ||
CX219686.1 | - | 2 - 3 | 183 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |