Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161111.1 | NP_001154583.1 | 3 - 4 | 456 | 62 | |
NM_172574.2 | NP_766162.2 | 3 - 4 | 456 | 62 | ||
GenBank mRNA | AK053171.1 | - | 3 - 4 | 405 | 62 | |
AK089230.1 | - | 3 - 4 | 365 | 62 | ||
AK150094.1 | - | 3 - 4 | 368 | 62 | ||
BC025220.1 | - | 3 - 4 | 394 | 5025905 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |