Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081090.1 | NP_001074559.1 | 7 - 8 | 1619 | 68 | |
GenBank mRNA | AK132230.1 | - | 6 - 7 | 1541 | 68 | |
BC010672.1 | - | 7 - 8 | 1590 | 68 | ||
BC021330.1 | - | 7 - 8 | 1638 | 68 | ||
BC038028.1 | - | 7 - 8 | 961 | 68 | ||
BC055464.1 | - | 6 - 7 | 1524 | 68 | ||
BC096029.1 | - | 7 - 8 | 1616 | 68 | ||
BC150893.1 | - | 7 - 8 | 1585 | 9087804 | 68 | |
BC157979.1 | - | 7 - 8 | 1585 | 9007299 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |