Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028816.2 | NP_083092.2 | 16 - 17 | 2656 | 55 | |
GenBank mRNA | AB093235.1 | - | 11 - 12 | 2921 | 55 | |
AK154709.1 | - | 16 - 17 | 2701 | 55 | ||
AK197184.1 | - | 1 - 2 | 88 | 55 | ||
AY029528.1 | - | 15 - 16 | 2152 | 55 | ||
BC058090.1 | - | 16 - 17 | 2656 | 6826356 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |