Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181729.2 | NP_859418.1 | 1260 | 104 | ||
GenBank mRNA | BC120907.2 | - | 9 - 10 | 1148 | 40087870 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |