Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290830.1 | NP_001277759.1 | 8 - 9 | 1077 | 75 | |
NM_013735.4 | NP_038763.3 | 8 - 9 | 1077 | 75 | ||
GenBank mRNA | AJ414734.1 | - | 7 - 8 | 938 | 75 | |
AK045851.1 | - | 8 - 9 | 992 | 75 | ||
AK148123.1 | - | 8 - 9 | 1037 | 75 | ||
BC079906.1 | - | 8 - 9 | 1011 | 30639375 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |