Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024432.2 | NP_077752.1 | 9 - 10 | 1121 | 63 | |
GenBank mRNA | AF272895.1 | - | 9 - 10 | 1136 | 63 | |
AK008942.1 | - | 9 - 10 | 1129 | 63 | ||
AK009117.1 | - | 8 - 9 | 903 | 63 | ||
AK075705.1 | - | 7 - 8 | 2185 | 63 | ||
AK082510.1 | - | 1160 | 63 | |||
AK131187.1 | - | 3 - 4 | 2893 | 63 | ||
AK148170.1 | - | 1089 | 63 | |||
AK161273.1 | - | 9 - 10 | 1142 | 63 | ||
BC016412.1 | - | 9 - 10 | 1050 | 4215883 | 63 | |
BC049963.1 | - | 9 - 10 | 1121 | 2655935 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |