Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uvrag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178635.3 | NP_848750.3 | 5 - 6 | 702 | 71 | |
GenBank mRNA | AB020334.1 | - | 4 - 5 | 307 | 71 | |
AK030814.1 | - | 5 - 6 | 633 | 71 | ||
AK043084.1 | - | 5 - 6 | 680 | 71 | ||
AK044129.1 | - | 6 - 7 | 798 | 71 | ||
AK078085.1 | - | 5 - 6 | 694 | 71 | ||
AK089900.1 | - | 5 - 6 | 687 | 71 | ||
AK154903.1 | - | 5 - 6 | 702 | 71 | ||
AK165015.1 | - | 5 - 6 | 72 | |||
AK165185.1 | - | 5 - 6 | 682 | 71 | ||
AK190073.1 | - | 67 | 71 | |||
AK215112.1 | - | 1 - 2 | 68 | 71 | ||
BC034176.1 | - | 5 - 6 | 671 | 4954871 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |