Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181664.2 | NP_858050.1 | 7 - 8 | 567 | 77 | |
GenBank mRNA | AF367970.1 | - | 7 - 8 | 567 | 77 | |
BC116360.1 | - | 7 - 8 | 567 | 40099973 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |