Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009358.3 | NP_033384.2 | 3 - 4 | 436 | 75 | |
GenBank mRNA | AB053464.1 | - | 3 - 4 | 381 | 75 | |
AF316616.2 | - | 3 - 4 | 293 | 75 | ||
AK154050.1 | - | 3 - 4 | 412 | 75 | ||
AK160587.1 | - | 3 - 4 | 377 | 75 | ||
AY338234.1 | - | 3 - 4 | 366 | 75 | ||
BC010716.1 | - | 3 - 4 | 349 | 2655861 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |