Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080575.2 | NP_542142.1 | 10 - 11 | 1593 | 69 | |
GenBank mRNA | AB046742.1 | - | 10 - 11 | 1582 | 69 | |
AK088244.1 | - | 10 - 11 | 1572 | 69 | ||
AK146568.1 | - | 10 - 11 | 1594 | 69 | ||
AK158119.1 | - | 10 - 11 | 1588 | 69 | ||
AK172470.1 | - | 10 - 11 | 1568 | 69 | ||
AK173275.1 | - | 10 - 11 | 1554 | 69 | ||
BC030930.1 | - | 6 - 7 | 767 | 4911679 | 69 | |
BC092278.1 | - | 10 - 11 | 1622 | 3710505 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |