Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fchsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199012.2 | NP_950177.2 | 11 - 12 | 1241 | 80 | |
GenBank mRNA | AK122368.1 | - | 11 - 12 | 1133 | 80 | |
BC060647.1 | - | 11 - 12 | 1203 | 6854041 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |