Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700032450.mm.1 |
NM_001252552.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032450.mm.1 |
NM_001252552.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032450.mm.1 |
NM_001252553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032450.mm.1 |
NM_001252553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032450.mm.1 |
NM_001252554.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032450.mm.1 |
NM_001252554.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032450.mm.1 |
NM_008034.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032450.mm.1 |
NM_008034.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032450.mm.1 |
TC0700003846.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032443.mm.1 |
NM_001252552.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032443.mm.1 |
NM_001252552.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032443.mm.1 |
NM_001252553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032443.mm.1 |
NM_001252553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032443.mm.1 |
NM_001252554.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032443.mm.1 |
NM_001252554.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032443.mm.1 |
NM_008034.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032443.mm.1 |
NM_008034.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032443.mm.1 |
TC0700003846.mm.1 |
MTA TC |