Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P512553 |
BP767155.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P512553 |
AK131109.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P512553 |
AK157410.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P512553 |
NM_001111111.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P512553 |
AK010773.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P512553 |
BP767155.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P512553 |
AK131109.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P512553 |
AK157410.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P512553 |
NM_001111111.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P512553 |
AK010773.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1985428 |
BC060054.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1985428 |
BP767155.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1985428 |
AK131109.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1985428 |
AK157410.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1985428 |
AK158186.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
292493 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
13571 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
833774 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
557860 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
332321 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
794181 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
315500 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
112641 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
869851 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
157605 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P116181 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P160721 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P512553 |
BP767155.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P512553 |
AK131109.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P512553 |
AK157410.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P512553 |
NM_001111111.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P512553 |
AK010773.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P512553 |
BP767155.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P512553 |
AK131109.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P512553 |
AK157410.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P512553 |
NM_001111111.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P512553 |
AK010773.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1985428 |
BC060054.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1985428 |
BP767155.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1985428 |
AK131109.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1985428 |
AK157410.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1985428 |
AK158186.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P512553 |
BP767155.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P512553 |
AK131109.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P512553 |
AK157410.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P512553 |
NM_001111111.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P512553 |
AK010773.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P512553 |
BP767155.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P512553 |
AK131109.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P512553 |
AK157410.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P512553 |
NM_001111111.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P512553 |
AK010773.1 |
mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372544 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372500 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372453 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372386 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372495 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372469 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372505 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372526 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372474 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372388 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372492 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372400 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372401 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372490 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372393 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372395 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372503 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372473 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372423 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25372399 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06529919 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372464 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06448533 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372533 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372447 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372474 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372516 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372542 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372386 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06529914 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372504 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06096714 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06262385 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372456 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372512 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372406 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372471 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372489 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06185222 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372442 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372415 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06262368 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372485 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06610236 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372422 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06125084 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372413 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06256664 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372382 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372450 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372404 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372424 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372451 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372493 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372501 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372460 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372416 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06338104 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372515 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06633381 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372531 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372539 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372508 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372492 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372469 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372462 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06493874 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372410 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06125085 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372523 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372529 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372459 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06351987 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372446 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372408 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372505 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372510 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06083110 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372383 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06060355 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372457 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372399 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372535 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372465 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372436 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372411 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372452 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372519 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06256672 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372537 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372466 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372423 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372499 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372434 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372454 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372472 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372402 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372486 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372431 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06351966 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372496 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06557268 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06296789 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06083108 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372393 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372498 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372384 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372536 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372487 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06433462 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372509 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372481 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06351974 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06610237 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372420 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06185219 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06529908 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06541744 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372513 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372407 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372525 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06514340 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06256666 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06622836 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372495 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25372401 |
- |
- |