Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163182.1 | NP_001156654.1 | 6 - 7 | 687 | 70 | |
NM_001163184.1 | NP_001156656.1 | 5 - 6 | 540 | 70 | ||
NM_001163185.1 | NP_001156657.1 | 5 - 6 | 530 | 70 | ||
NM_001163187.1 | NP_001156659.1 | 4 - 5 | 531 | 70 | ||
GenBank mRNA | AF071000.1 | - | 4 - 5 | 519 | 70 | |
AF101053.1 | - | 6 - 7 | 658 | 70 | ||
AK005102.1 | - | 5 - 6 | 495 | 70 | ||
AK141634.1 | - | 498 | 70 | |||
AK158172.1 | - | 5 - 6 | 531 | 70 | ||
AK164683.1 | - | 5 - 6 | 495 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |