Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177324.2 | NP_796298.2 | 10 - 11 | 1213 | 83 | |
GenBank mRNA | AK046622.1 | - | 10 - 11 | 1201 | 83 | |
AK048952.1 | - | 10 - 11 | 1190 | 83 | ||
AK080952.1 | - | 10 - 11 | 1214 | 83 | ||
AK080963.1 | - | 10 - 11 | 1208 | 83 | ||
AK087396.1 | - | 10 - 11 | 1208 | 83 | ||
AK173257.1 | - | 10 - 11 | 1172 | 83 | ||
BC048706.1 | - | 10 - 11 | 1198 | 83 | ||
BC067204.1 | - | 11 - 12 | 1251 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |