Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020575.2 | NP_065600.1 | 5 - 6 | 1776 | 83 | |
GenBank mRNA | AF155739.1 | - | 5 - 6 | 1776 | 83 | |
AK034109.1 | - | 1766 | 83 | |||
AK166666.1 | - | 5 - 6 | 1783 | 83 | ||
AK167246.1 | - | 5 - 6 | 1758 | 83 | ||
AK168555.1 | - | 5 - 6 | 1779 | 83 | ||
BC025029.1 | - | 5 - 6 | 1740 | 4935124 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |