Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK044224.1 | - | 3 - 4 | 303 | 72 | |
AK053879.1 | - | 3 - 4 | 339 | 72 | ||
AK083340.1 | - | 3 - 4 | 325 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |