Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133942.2 | NP_598703.1 | 6 - 7 | 638 | 135 | |
GenBank mRNA | AK044224.1 | - | 8 - 9 | 790 | 135 | |
AK083340.1 | - | 8 - 9 | 812 | 135 | ||
BC020017.1 | - | 6 - 7 | 638 | 3589001 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |