Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159646.1 | NP_001153118.1 | 2 - 3 | 285 | 133 | |
NM_023595.6 | NP_076084.2 | 1 - 2 | 380 | 133 | ||
GenBank mRNA | AF091101.1 | - | 1 - 2 | 237 | 133 | |
AK003076.1 | - | 1 - 2 | 169 | 133 | ||
AK011407.1 | - | 1 - 2 | 171 | 133 | ||
AK017216.1 | - | 2 - 3 | 275 | 133 | ||
AK077244.1 | - | 1 - 2 | 413 | 133 | ||
AK135438.1 | - | 2 - 3 | 294 | 133 | ||
AK152459.1 | - | 1 - 2 | 176 | 133 | ||
AK153285.1 | - | 1 - 2 | 161 | 133 | ||
BC019979.1 | - | 2 - 3 | 267 | 4187559 | 133 | |
BC053693.1 | - | 1 - 2 | 167 | 6485182 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |