Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030691.1 | NP_109616.1 | 2 - 3 | 448 | 67 | |
GenBank mRNA | AJ278575.1 | - | 2 - 3 | 448 | 67 | |
AK040569.1 | - | 2 - 3 | 428 | 67 | ||
AK151417.1 | - | 2 - 3 | 437 | 67 | ||
AK152186.1 | - | 2 - 3 | 437 | 67 | ||
AK156382.1 | - | 2 - 3 | 386 | 67 | ||
AK163058.1 | - | 2 - 3 | 428 | 67 | ||
BC057951.1 | - | 2 - 3 | 445 | 5341788 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |