Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143765.1 | NP_001137237.1 | 7 - 8 | 510 | 123 | |
GenBank mRNA | AK016694.1 | - | 7 - 8 | 510 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |