Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178755.3 | NP_848870.2 | 3 - 4 | 262 | 107 | |
GenBank mRNA | AK029744.1 | - | 3 - 4 | 295 | 107 | |
AK029932.1 | - | 3 - 4 | 263 | 107 | ||
DQ867026.1 | - | 3 - 4 | 295 | 107 | ||
DQ867027.1 | - | 3 - 4 | 295 | 107 | ||
DQ867028.1 | - | 3 - 4 | 295 | 107 | ||
DQ867029.1 | - | 3 - 4 | 295 | 107 | ||
DQ867030.1 | - | 3 - 4 | 295 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |