Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178890.3 | NP_849221.2 | 15 - 16 | 3179 | 64 | |
GenBank mRNA | AK042322.1 | - | 15 - 16 | 3179 | 64 | |
AK151620.1 | - | 15 - 16 | 3175 | 64 | ||
AK152163.1 | - | 15 - 16 | 3175 | 64 | ||
AK170569.1 | - | 15 - 16 | 3178 | 64 | ||
AK172414.1 | - | 15 - 16 | 3177 | 64 | ||
AK172468.1 | - | 15 - 16 | 3176 | 64 | ||
BC006756.1 | - | 6 - 7 | 719 | 64 | ||
BC040825.1 | - | 1 - 2 | 490 | 64 | ||
BC054399.1 | - | 15 - 16 | 3179 | 6493840 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |