Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpine2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009255.4 | NP_033281.1 | 6 - 7 | 1159 | 127 | |
GenBank mRNA | AK045954.1 | - | 6 - 7 | 1131 | 127 | |
AK047856.1 | - | 6 - 7 | 1131 | 127 | ||
AK051332.1 | - | 6 - 7 | 1131 | 127 | ||
AK159687.1 | - | 6 - 7 | 1131 | 127 | ||
X70296.1 | - | 6 - 7 | 1130 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |