Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010576.3 | NP_034706.3 | 9 - 10 | 1280 | 73 | |
GenBank mRNA | AB850880.1 | - | 9 - 10 | 1045 | 73 | |
AF109136.1 | - | 9 - 10 | 1116 | 73 | ||
AK051676.1 | - | 9 - 10 | 1206 | 73 | ||
AK089898.1 | - | 8 - 9 | 1080 | 73 | ||
AK156015.1 | - | 9 - 10 | 1176 | 73 | ||
AK156517.1 | - | 9 - 10 | 1050 | 73 | ||
BC068313.1 | - | 9 - 10 | 1060 | 30093248 | 73 | |
X53176.1 | - | 9 - 10 | 1203 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |