Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007389.5 | NP_031415.2 | 7 - 8 | 1044 | 95 | |
GenBank mRNA | AK029177.1 | - | 7 - 8 | 1013 | 95 | |
AK132589.1 | - | 7 - 8 | 984 | 95 | ||
BC125439.1 | - | 7 - 8 | 1044 | 40129954 | 95 | |
BC137678.1 | - | 7 - 8 | 1044 | 8860697 | 95 | |
X03986.1 | - | 7 - 8 | 1001 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |