Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144887.4 | NP_659136.1 | 7 - 8 | 1985 | 87 | |
GenBank mRNA | AK028737.1 | - | 4 - 5 | 551 | 87 | |
AK082513.1 | - | 7 - 8 | 1910 | 87 | ||
AK145086.1 | - | 7 - 8 | 1912 | 87 | ||
AK173251.1 | - | 7 - 8 | 1940 | 87 | ||
AY894891.1 | - | 7 - 8 | 1977 | 87 | ||
BC020051.1 | - | 6 - 7 | 955 | 3987696 | 87 | |
BC027047.1 | - | 3 - 4 | 219 | 87 | ||
BC065155.1 | - | 7 - 8 | 1855 | 5706087 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |