Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700009299.mm.1 |
NM_025887.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009299.mm.1 |
NM_025887.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009299.mm.1 |
TC1700000973.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009300.mm.1 |
NM_025887.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009300.mm.1 |
NM_025887.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009300.mm.1 |
TC1700000973.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009298.mm.1 |
NM_025887.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009298.mm.1 |
NM_025887.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009298.mm.1 |
TC1700000973.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009302.mm.1 |
NM_025887.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009302.mm.1 |
NM_025887.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009302.mm.1 |
TC1700000973.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009304.mm.1 |
NM_025887.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009304.mm.1 |
NM_025887.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009304.mm.1 |
TC1700000973.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009305.mm.1 |
NM_025887.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009305.mm.1 |
NM_025887.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009305.mm.1 |
TC1700000973.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009303.mm.1 |
NM_025887.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009303.mm.1 |
NM_025887.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009303.mm.1 |
TC1700000973.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009301.mm.1 |
NM_025887.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009301.mm.1 |
NM_025887.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009301.mm.1 |
TC1700000973.mm.1 |
MTA TC |