Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mki67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081117.2 | NP_001074586.2 | 1 - 2 | 327 | 82 | |
GenBank mRNA | AK052772.1 | - | 1 - 2 | 247 | 82 | |
AK082099.1 | - | 1 - 2 | 245 | 82 | ||
AK143677.1 | - | 1 - 2 | 366 | 82 | ||
X82786.1 | - | 1 - 2 | 432 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |