Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033317.3 | NP_001028489.1 | 1 - 2 | 182 | 110 | |
GenBank mRNA | AK136723.1 | - | 1 - 2 | 184 | 110 | |
BC107347.1 | - | 1 - 2 | 128 | 40055406 | 110 | |
BC107348.2 | - | 1 - 2 | 128 | 40055413 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |