Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009844.2 | NP_033974.2 | 8 - 9 | 1214 | 75 | |
GenBank mRNA | AK089835.1 | - | 8 - 9 | 1213 | 75 | |
AK156125.1 | - | 8 - 9 | 1214 | 75 | ||
AK163608.1 | - | 6 - 7 | 1711 | 75 | ||
AK172008.1 | - | 8 - 9 | 1176 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |