Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287171.1 | NP_001274100.1 | 2 - 3 | 222 | 102 | |
NM_011328.3 | NP_035458.1 | 2 - 3 | 222 | 102 | ||
GenBank mRNA | AK145859.1 | - | 2 - 3 | 221 | 102 | |
AK167408.1 | - | 2 - 3 | 221 | 102 | ||
BC048484.1 | - | 2 - 3 | 222 | 6707701 | 102 | |
X73580.1 | - | 2 - 3 | 195 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |