Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuzd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008411.3 | NP_032437.3 | 2 - 3 | 482 | 113 | |
GenBank mRNA | BC140992.1 | - | 2 - 3 | 417 | 9055604 | 113 |
BY707834.1 | - | 2 - 3 | 248 | 113 | ||
U69699.2 | - | 2 - 3 | 484 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |