Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009627.1 | NP_033757.1 | 2 - 3 | 272 | 65 | |
GenBank mRNA | AK144181.1 | - | 2 - 3 | 287 | 65 | |
AK144600.1 | - | 2 - 3 | 281 | 65 | ||
AK144649.1 | - | 2 - 3 | 285 | 65 | ||
BC052665.1 | - | 2 - 3 | 282 | 30054008 | 65 | |
U77630.1 | - | 2 - 3 | 272 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons |