Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB036992.1 | - | 9 - 10 | 1496 | 112 | |
AK089975.1 | - | 9 - 10 | 1474 | 112 | ||
AK090084.1 | - | 9 - 10 | 1550 | 112 | ||
BC057890.1 | - | 9 - 10 | 1491 | 4979982 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |