Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145488.1 | NP_663463.1 | 12 - 13 | 2406 | 94 | |
GenBank mRNA | BC003424.1 | - | 12 - 13 | 2406 | 2609884 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |