Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111142.1 | NP_001104612.1 | 3 - 4 | 737 | 70 | |
GenBank mRNA | AK134769.1 | - | 1 - 2 | 1803 | 70 | |
BC147714.1 | - | 3 - 4 | 712 | 9087473 | 70 | |
BC172136.1 | - | 3 - 4 | 71 |
Set Membership: |
Amplicon length less than or equal to 70 Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |