Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486499 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486500 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486497 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486498 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486501 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486504 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486502 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2083988 |
BC057880.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2083988 |
BX523983.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2083988 |
NM_001142335.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2083988 |
NM_001142336.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2083988 |
NM_001142337.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
115085 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
59285 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354852 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
30806 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
609169 |
AK005571.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
609169 |
BC059727.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
609169 |
NM_011449.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
609169 |
Z46299.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
282815 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
526288 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
15851 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
70769 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
201039 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
474003 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
880845 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
816469 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438641 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
99159 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
95916 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P162116 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2083988 |
BC057880.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2083988 |
BX523983.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2083988 |
NM_001142335.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2083988 |
NM_001142336.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2083988 |
NM_001142337.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P223686 |
M64360.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P223686 |
W76814.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P223686 |
AK013416.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P223686 |
AK217345.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P223686 |
BC057880.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486467 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486483 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486493 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486461 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486512 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486472 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486490 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486502 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486475 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486513 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486517 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486486 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486470 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486515 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486495 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486487 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486460 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486480 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486510 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486496 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486466 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486469 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486514 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486462 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486499 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486474 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486464 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486504 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486478 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486485 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486508 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21486463 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P162116 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00449224 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P223686 |
M64360.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P223686 |
W76814.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P223686 |
AK013416.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P223686 |
AK217345.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P223686 |
BC057880.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P131230 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108844 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P00449224 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P162116 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01994133 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01325925 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01994130 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02258236 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01994128 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486497 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486499 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486502 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486500 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486498 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486504 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21486501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486469 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486558 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486543 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486619 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486495 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486455 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486584 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486475 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486549 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486526 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486534 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486597 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486490 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486571 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486559 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486554 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486530 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21486486 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486480 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486588 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01356955 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486475 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486566 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486587 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486626 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486446 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486467 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486559 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486521 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486523 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486522 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486478 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486502 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486546 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486439 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01460829 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01625003 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486437 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486526 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486556 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486515 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486570 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486504 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486449 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486537 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01750051 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486598 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01737257 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01325925 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486532 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486483 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486623 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486497 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486513 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486567 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486458 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486561 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486490 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486622 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486540 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486535 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01962911 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486514 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486601 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486621 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01994124 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02068173 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486527 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486538 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486545 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486633 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486453 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486533 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486584 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486597 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486518 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02068159 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01962910 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486487 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486593 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01636541 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486442 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486534 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01323259 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486627 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486512 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486571 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486624 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01750050 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02068154 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486558 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486460 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486594 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486486 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486634 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486510 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486443 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01603359 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486543 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02102931 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486485 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01612742 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01612744 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01935772 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486550 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486539 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486530 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486569 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01323255 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486455 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01994128 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02068167 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02068175 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01323254 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486508 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01603365 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486590 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486450 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01356952 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01323257 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486547 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486579 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486472 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01737277 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21486564 |
- |
- |