Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177660.3 | NP_808328.2 | 1 - 2 | 1511 | 65 | |
GenBank mRNA | CB525445.1 | - | 1 - 2 | 493 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus Inventoried Probe spans exons |