Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025285.2 | NP_079561.1 | 1 - 2 | 104 | 68 | |
GenBank mRNA | AK003659.1 | - | 1 - 2 | 104 | 68 | |
AK013020.1 | - | 1 - 2 | 105 | 68 | ||
AK149212.1 | - | 1 - 2 | 129 | 68 | ||
BC026538.1 | - | 1 - 2 | 66 | 4981703 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |