Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200013353.mm.1 |
NM_001172104.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013353.mm.1 |
NM_001172104.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013353.mm.1 |
NM_028356.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013353.mm.1 |
NM_028356.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013353.mm.1 |
TC1200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013352.mm.1 |
NM_001172104.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013352.mm.1 |
NM_001172104.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013352.mm.1 |
NM_028356.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013352.mm.1 |
NM_028356.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013352.mm.1 |
TC1200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013360.mm.1 |
NM_001172104.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013360.mm.1 |
NM_001172104.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013360.mm.1 |
TC1200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013361.mm.1 |
NM_001172104.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013361.mm.1 |
NM_001172104.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013361.mm.1 |
TC1200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013354.mm.1 |
NM_001172104.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013354.mm.1 |
NM_001172104.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013354.mm.1 |
NM_028356.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013354.mm.1 |
NM_028356.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013354.mm.1 |
TC1200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013355.mm.1 |
NM_001172104.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013355.mm.1 |
NM_001172104.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013355.mm.1 |
NM_028356.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013355.mm.1 |
NM_028356.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013355.mm.1 |
TC1200001947.mm.1 |
MTA TC |