Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012024.2 | NP_036154.1 | 5 - 6 | 1120 | 80 | |
GenBank mRNA | AK088366.1 | - | 5 - 6 | 1092 | 80 | |
AK132432.1 | - | 5 - 6 | 1153 | 80 | ||
AK133824.1 | - | 4 - 5 | 1063 | 80 | ||
AK220163.1 | - | 5 - 6 | 1110 | 80 | ||
BC085149.1 | - | 5 - 6 | 1120 | 6848033 | 80 | |
BC094387.1 | - | 5 - 6 | 1112 | 80 | ||
U49728.1 | - | 3 - 4 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |