Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028772.3 | NP_083048.1 | 7 - 8 | 1185 | 97 | |
GenBank mRNA | AK004755.1 | - | 7 - 8 | 1186 | 97 | |
BC024126.1 | - | 3 - 4 | 468 | 97 | ||
BC089599.1 | - | 7 - 8 | 1188 | 30118948 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |