Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10605048 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605025 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605021 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605022 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605045 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605022 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605025 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605047 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605019 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605050 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605024 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605027 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605026 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605027 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605046 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605046 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605049 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605048 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605020 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605024 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605023 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605023 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605021 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605047 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605045 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605019 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605026 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605049 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10605044 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10605018 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10605018 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10605044 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00014929.mm.1 |
NM_001045539.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014929.mm.1 |
NM_001045539.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014929.mm.1 |
TC0X00003427.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004259.mm.1 |
NM_001111293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004259.mm.1 |
NM_001111293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004259.mm.1 |
TC0X00000675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004261.mm.1 |
NM_001111293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004261.mm.1 |
NM_001111293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004261.mm.1 |
TC0X00000675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014928.mm.1 |
NM_001045539.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014928.mm.1 |
NM_001045539.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014928.mm.1 |
TC0X00003427.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004260.mm.1 |
NM_001111293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004260.mm.1 |
NM_001111293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004260.mm.1 |
TC0X00000675.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1422933_PM_at |
- |
- |
Mouse Expression Set 430 |
1422933_PM_at |
- |
- |
Mouse Expression Set 430 |
1422933_at |
- |
- |
Mouse Expression Set 430 |
1422933_at |
- |
- |
Mouse Expression Set 430 |
1443483_at |
- |
- |
Mouse Expression Set 430 |
1443483_PM_at |
- |
- |
Mouse Expression Set 430 |
1443483_PM_at |
- |
- |
Mouse Expression Set 430 |
1443483_at |
- |
- |
Murine Genome U74v2 Set |
133850_at |
- |
- |
Murine Genome U74v2 Set |
133850_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1422933_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1422933_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1422933_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1422933_PM_at |
- |
- |